siRNA Details
| Gene Symbol: SDK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144952.1 | NP_001138424.1 | 26, 27 | 3702 | ||
| XM_011524914.2 | XP_011523216.1 | 26, 27 | 4086 | |||
| XM_011524915.2 | XP_011523217.1 | 26, 27 | 4088 | |||
| GenBank mRNA | AB040947.1 | - | 19, 20 | 2733 | ||
| AL137357.1 | - | 4, 5 | 610 | |||
| BC045763.1 | - | 8, 9 | 1230 | |||
| BC066363.1 | - | 18, 19 | 2602 | |||

SDK2
Genomic Map