siRNA Details
| Gene Symbol: CRLS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127458.1 | NP_001120930.1 | 3, 4 | 434 | ||
| NM_001323561.1 | NP_001310490.1 | 3, 4 | 442 | |||
| NM_001323562.1 | NP_001310491.1 | 3, 4 | 449 | |||
| NM_001323563.1 | NP_001310492.1 | 3, 4 | 817 | |||
| NM_001323564.1 | NP_001310493.1 | 3, 4 | 443 | |||
| NM_019095.5 | NP_061968.1 | 3, 4 | 755 | |||
| XM_005260738.4 | XP_005260795.1 | 3, 4 | 419 | |||
| XM_011529263.1 | XP_011527565.1 | 4, 5 | 773 | |||
| GenBank mRNA | AF241784.1 | - | 3, 4 | 674 | ||
| AL832419.1 | - | 3, 4 | 502 | |||
| BM563835.1 | - | 3, 4 | 437 | |||
| BP368984.1 | - | 3, 4 | 434 | |||
| BU852955.1 | - | 4, 5 | 462 | |||
| DQ386730.1 | - | 3, 4 | 581 | |||

CRLS1
Genomic Map