siRNA Details
| Gene Symbol: PCSK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017573.4 | NP_060043.2 | 15 | 2291 | ||
| XM_005259586.1 | XP_005259643.1 | 14 | 2044 | |||
| XM_011528085.2 | XP_011526387.1 | 16 | 2516 | |||
| XM_011528086.2 | XP_011526388.1 | 16 | 2489 | |||
| XM_011528087.2 | XP_011526389.1 | 15 | 2342 | |||
| XM_011528088.2 | XP_011526390.1 | 16 | 2168 | |||
| XM_011528089.2 | XP_011526391.1 | 14 | 2234 | |||
| XM_011528090.2 | XP_011526392.1 | 13 | 2153 | |||
| XM_011528091.2 | XP_011526393.1 | 14 | 1989 | |||
| XM_011528093.2 | XP_011526395.1 | 15 | 2391 | |||
| XM_011528094.1 | XP_011526396.1 | 13 | 2123 | |||
| XM_011528096.1 | XP_011526398.1 | 13 | 1851 | |||
| GenBank mRNA | AK057235.1 | - | 12 | 2229 | ||
| AL133566.1 | - | 3 | 613 | |||
| AY358963.1 | - | 15 | 2249 | |||
| BC036354.1 | - | 5 | 991 | |||
| BC142942.1 | - | 10 | 2442 | |||
| BU742489.1 | - | 1 | 427 | |||

PCSK4
Genomic Map