siRNA Details
| Gene Symbol: BNC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317939.1 | NP_001304868.1 | 5 | 2217 | ||
| NM_001317940.1 | NP_001304869.1 | 5 | 2123 | |||
| NM_017637.5 | NP_060107.3 | 6 | 2343 | |||
| XM_011517924.1 | XP_011516226.1 | 5 | 2267 | |||
| XM_011517934.2 | XP_011516236.1 | 3 | 3295 | |||
| XM_017014816.1 | XP_016870305.1 | 8 | 4653 | |||
| XM_017014817.1 | XP_016870306.1 | 7 | 2802 | |||
| XM_017014818.1 | XP_016870307.1 | 6 | 2717 | |||
| XM_017014819.1 | XP_016870308.1 | 7 | 2801 | |||
| XM_017014820.1 | XP_016870309.1 | 7 | 2801 | |||
| XM_017014821.1 | XP_016870310.1 | 6 | 2715 | |||
| XM_017014822.1 | XP_016870311.1 | 4 | 2263 | |||
| XM_017014823.1 | XP_016870312.1 | 4 | 7032 | |||
| XM_017014824.1 | XP_016870313.1 | 3 | 2584 | |||
| XM_017014825.1 | XP_016870314.1 | 7 | 2800 | |||
| XM_017014826.1 | XP_016870315.1 | 7 | 2800 | |||
| XM_017014827.1 | XP_016870316.1 | 7 | 2800 | |||
| XM_017014828.1 | XP_016870317.1 | 7 | 2799 | |||
| XM_017014829.1 | XP_016870318.1 | 5 | 2771 | |||
| GenBank mRNA | AK000050.1 | - | 1 | 127 | ||
| AK092247.1 | - | 1 | 1073 | |||
| AK131398.1 | - | 6 | 2225 | |||
| AK299073.1 | - | 5 | 2093 | |||
| AK304745.1 | - | 3 | 1885 | |||
| AY438376.1 | - | 4 | 1988 | |||
| BX641030.1 | - | 1 | 1373 | |||
| CR933649.1 | - | 5 | 2174 | |||

BNC2
Genomic Map