siRNA Details
| Gene Symbol: AFTPH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002243.2 | NP_001002243.1 | 2 | 902 | ||
| NM_017657.4 | NP_060127.3 | 2 | 902 | |||
| NM_203437.3 | NP_982261.2 | 2 | 902 | |||
| XM_005264380.1 | XP_005264437.1 | 2 | 902 | |||
| GenBank mRNA | AB073356.1 | - | 1 | 344 | ||
| AK000087.1 | - | 2 | 877 | |||
| AK222821.1 | - | 2 | 825 | |||
| AL833962.2 | - | 2 | 680 | |||
| AY367088.1 | - | 1 | 562 | |||
| BC047529.1 | - | 2 | 876 | 5311682 | ||

AFTPH
Genomic Map