siRNA Details
| Gene Symbol: QPCTL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163377.1 | NP_001156849.1 | 2 | 498 | ||
| NM_017659.3 | NP_060129.2 | 2 | 498 | |||
| XM_011527048.2 | XP_011525350.1 | 2 | 579 | |||
| XM_017026900.1 | XP_016882389.1 | 2 | 577 | |||
| GenBank mRNA | AK000091.1 | - | 2 | 307 | ||
| AK172764.1 | - | 2 | 306 | |||
| AK222636.1 | - | 2 | 291 | |||
| BC011553.1 | - | 2 | 284 | 4564051 | ||
| BP216511.1 | - | 2 | 499 | |||

QPCTL
Genomic Map