siRNA Details
| Gene Symbol: PGPEP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300927.1 | NP_001287856.1 | 2 | 159 | ||
| NM_001308366.1 | NP_001295295.1 | 2 | 159 | |||
| NM_017712.3 | NP_060182.1 | 2 | 159 | |||
| XM_006722783.3 | XP_006722846.1 | 2 | 173 | |||
| XM_011528111.2 | XP_011526413.1 | 2 | 159 | |||
| XM_011528112.1 | XP_011526414.1 | 2 | 159 | |||
| GenBank mRNA | AJ278828.1 | - | 2 | 106 | ||
| AK000215.1 | - | 2 | 106 | |||
| AK289968.1 | - | 2 | 159 | |||
| BC028063.1 | - | 2 | 132 | |||
| BC042138.1 | - | 2 | 135 | 5935845 | ||
| BX473633.1 | - | 2 | 159 | |||
| CF129240.1 | - | 2 | 86 | |||
| DA315740.1 | - | 2 | 159 | |||
| DA738481.1 | - | 2 | 148 | |||
| DB276426.1 | - | 2 | 109 | |||
| Gene Symbol: FKBP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK026952.1 | - | 103 | |||

PGPEP1
Genomic Map