siRNA Details
| Gene Symbol: PALMD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017734.4 | NP_060204.1 | 7 | 1159 | ||
| GenBank mRNA | AF262379.1 | - | 7 | 1090 | ||
| AJ312214.1 | - | 7 | 1001 | |||
| AK000278.1 | - | 7 | 921 | |||
| AK024654.1 | - | 2 | 222 | |||
| AL390970.1 | - | 1 | 188 | |||
| AL390972.1 | - | 7 | 598 | |||
| AL390973.1 | - | 7 | 885 | |||
| BC012570.1 | - | 7 | 974 | 4245261 | ||
| BX648213.1 | - | 4 | 3874 | |||
| KU178657.1 | - | 7 | 716 | |||

PALMD
Genomic Map