siRNA Details
| Gene Symbol: PLPPR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017753.2 | NP_060223.2 | 4 | 632 | ||
| NM_207299.1 | NP_997182.1 | 4 | 765 | |||
| GenBank mRNA | AK000307.1 | - | 4 | 632 | ||
| AK225306.1 | - | 4 | 632 | |||
| AK303718.1 | - | 2 | 512 | |||
| AK303807.1 | - | 3 | 628 | |||
| AK309115.1 | - | 2 | 341 | |||
| AY304515.1 | - | 4 | 498 | |||
| AY337718.1 | - | 3 | 378 | |||
| BC022465.1 | - | 4 | 498 | 4797229 | ||
| CA772610.1 | - | 4 | 668 | |||

PLPPR1
Genomic Map