siRNA Details
| Gene Symbol: RNF43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001305544.1 | NP_001292473.1 | 9 | 2663 | ||
| NM_001305545.1 | NP_001292474.1 | 8 | 1745 | |||
| NM_017763.5 | NP_060233.3 | 9 | 2866 | |||
| XM_011524955.2 | XP_011523257.1 | 9 | 2765 | |||
| XM_011524956.2 | XP_011523258.1 | 8 | 3542 | |||
| XM_017024800.1 | XP_016880289.1 | 10 | 4092 | |||
| GenBank mRNA | AB081837.1 | - | 9 | 2403 | ||
| AK000322.1 | - | 8 | 2083 | |||
| AK291027.1 | - | 9 | 2636 | |||
| AK296769.1 | - | 7 | 1856 | |||
| AK298789.1 | - | 4 | 1276 | |||
| AK299024.1 | - | 8 | 1745 | |||
| AK309297.1 | - | 8 | 1732 | |||
| BC109028.2 | - | 8 | 2112 | 40027722 | ||
| CR627423.1 | - | 9 | 2663 | |||
| CR749257.1 | - | 8 | 3871 | |||

RNF43
Genomic Map