siRNA Details
| Gene Symbol: WHSC1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017778.2 | NP_060248.2 | 2 | 961 | ||
| NM_023034.1 | NP_075447.1 | 2 | 961 | |||
| GenBank mRNA | AF255649.1 | - | 3 | 560 | ||
| AF318339.1 | - | 2 | 1132 | |||
| AF332468.1 | - | 2 | 961 | |||
| AF332469.1 | - | 2 | 961 | |||
| AJ295990.1 | - | 2 | 756 | |||
| AJ295991.1 | - | 2 | 756 | |||
| AJ295992.1 | - | 2 | 756 | |||
| AK022560.1 | - | 2 | 638 | |||
| AK127594.1 | - | 3 | 1481 | |||
| BC012059.1 | - | 2 | 929 | |||
| BC062631.1 | - | 2 | 900 | |||
| BC073858.1 | - | 1 | 487 | |||
| BC101717.1 | - | 2 | 535 | 8069223 | ||
| BC107734.1 | - | 1 | 937 | |||
| BC113469.1 | - | 2 | 535 | 8322521 | ||
| BC115006.1 | - | 2 | 556 | 7961962 | ||
| BC143510.1 | - | 2 | 535 | 9052018 | ||

WHSC1L1
Genomic Map