siRNA Details
| Gene Symbol: FOCAD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017794.4 | NP_060264.4 | 6, 7 | 657 | ||
| XM_005251494.3 | XP_005251551.2 | 4, 5 | 641 | |||
| XM_017014852.1 | XP_016870341.1 | 5, 6 | 431 | |||
| XM_017014853.1 | XP_016870342.1 | 5, 6 | 431 | |||
| XM_017014855.1 | XP_016870344.1 | 5, 6 | 530 | |||
| XM_017014856.1 | XP_016870345.1 | 5, 6 | 471 | |||
| XM_017014857.1 | XP_016870346.1 | 4, 5 | 641 | |||
| GenBank mRNA | AK056522.1 | - | 6, 7 | 657 | ||
| AY139834.1 | - | 6, 7 | 656 | |||

FOCAD
Genomic Map