siRNA Details
| Gene Symbol: CHTF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039690.3 | NP_001034779.1 | 4 | 825 | ||
| NM_001040146.3 | NP_001035236.1 | 4 | 798 | |||
| NR_033227.1 | 3 | 767 | ||||
| XM_011523167.1 | XP_011521469.1 | 3 | 704 | |||
| XM_011523168.1 | XP_011521470.1 | 3 | 658 | |||
| GenBank mRNA | AF525164.1 | - | 4 | 805 | ||
| AK000407.1 | - | 4 | 805 | |||
| AL833902.1 | - | 1 | 384 | |||
| BC018700.2 | - | 4 | 772 | 3504183 | ||
| BC069059.1 | - | 3 | 688 | |||

CHTF8
Genomic Map