siRNA Details
| Gene Symbol: HAUS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166269.1 | NP_001159741.1 | 7 | 937 | ||
| NM_001166270.1 | NP_001159742.1 | 6 | 794 | |||
| NM_017815.2 | NP_060285.2 | 7 | 929 | |||
| GenBank mRNA | AK000431.1 | - | 7 | 898 | ||
| AK026016.1 | - | 7 | 883 | |||
| AY927599.1 | - | 2 | 177 | |||
| BC001916.1 | - | 7 | 926 | 2905474 | ||
| BC002554.2 | - | 7 | 860 | 3139198 | ||
| BX247995.1 | - | 6 | 746 | |||
| BX248256.1 | - | 6 | 761 | |||

HAUS4
Genomic Map