siRNA Details
| Gene Symbol: FAM120C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300788.1 | NP_001287717.1 | 6 | 1387 | ||
| NM_017848.5 | NP_060318.4 | 6 | 1364 | |||
| XM_006724589.3 | XP_006724652.1 | 6 | 1360 | |||
| GenBank mRNA | AY150025.1 | - | 6 | 1364 | ||
| BC136413.1 | - | 6 | 1361 | 9020401 | ||
| BC144081.1 | - | 6 | 1361 | |||

FAM120C
Genomic Map