siRNA Details
| Gene Symbol: C1orf123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017887.2 | NP_060357.1 | 2, 3 | 144 | ||
| GenBank mRNA | AK000587.1 | - | 2, 3 | 122 | ||
| BC010908.1 | - | 2, 3 | 126 | 4093763 | ||
| CR457246.1 | - | 1, 2 | 80 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C1orf123| Gene Symbol: C1orf123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017887.2 | NP_060357.1 | 2, 3 | 144 | ||
| GenBank mRNA | AK000587.1 | - | 2, 3 | 122 | ||
| BC010908.1 | - | 2, 3 | 126 | 4093763 | ||
| CR457246.1 | - | 1, 2 | 80 | |||