siRNA Details
| Gene Symbol: C1orf123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017887.2 | NP_060357.1 | 3 | 174 | ||
| GenBank mRNA | AK000587.1 | - | 3 | 152 | ||
| AK092734.1 | - | 1 | 1018 | |||
| BC010908.1 | - | 3 | 156 | 4093763 | ||
| CR457246.1 | - | 2 | 110 | |||

C1orf123
Genomic Map