siRNA Details
| Gene Symbol: ZNF770 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014106.3 | NP_054825.2 | 3 | 1507 | ||
| XM_011521744.2 | XP_011520046.1 | 2 | 1351 | |||
| GenBank mRNA | AI421027.1 | - | 1 | 91 | ||
| AK131433.1 | - | 1 | 379 | |||
| AK291438.1 | - | 3 | 1498 | |||
| BC071603.1 | - | 3 | 1479 | 30340520 | ||
| BM454360.1 | - | 1 | 28 | |||

ZNF770
Genomic Map