siRNA Details
| Gene Symbol: ZNF770 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014106.3 | NP_054825.2 | 3 | 2416 | ||
| XM_011521744.2 | XP_011520046.1 | 2 | 2260 | |||
| GenBank mRNA | AK127201.1 | - | 1 | 527 | ||
| AK131433.1 | - | 1 | 1288 | |||
| AK291438.1 | - | 3 | 2407 | |||
| BC071603.1 | - | 3 | 2388 | 30340520 | ||

ZNF770
Genomic Map