siRNA Details
| Gene Symbol: TESC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017899.3 | NP_060369.3 | 2, 3 | 286 | ||
| NR_031766.2 | 2, 3 | 286 | ||||
| XM_011538503.1 | XP_011536805.1 | 2, 3 | 101 | |||
| XM_017019532.1 | XP_016875021.1 | 2, 3 | 330 | |||
| XM_017019533.1 | XP_016875022.1 | 2, 3 | 330 | |||
| GenBank mRNA | AF443207.1 | - | 2, 3 | 243 | ||
| AK000614.1 | - | 2, 3 | 329 | |||
| BC015221.1 | - | 2, 3 | 243 | 3885280 | ||
| BQ434057.1 | - | 2, 3 | 219 | |||
| DB452909.1 | - | 2, 3 | 286 | |||

TESC
Genomic Map