siRNA Details
| Gene Symbol: TUG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002323.2 | 1 | 2545 | |||
| NR_110492.1 | 1 | 2545 | ||||
| NR_110493.1 | 1 | 2545 | ||||
| GenBank mRNA | AK000749.1 | - | 1 | 389 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TUG1| Gene Symbol: TUG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002323.2 | 1 | 2545 | |||
| NR_110492.1 | 1 | 2545 | ||||
| NR_110493.1 | 1 | 2545 | ||||
| GenBank mRNA | AK000749.1 | - | 1 | 389 | ||