siRNA Details
| Gene Symbol: TTC38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017931.2 | NP_060401.2 | 4 | 419 | ||
| XM_011530259.1 | XP_011528561.1 | 5 | 547 | |||
| XM_011530260.2 | XP_011528562.1 | 4 | 419 | |||
| GenBank mRNA | AK000706.1 | - | 4 | 375 | ||
| AK300606.1 | - | 4 | 399 | |||
| BC018918.2 | - | 4 | 360 | 4130906 | ||
| DA762669.1 | - | 4 | 421 | |||

TTC38
Genomic Map