siRNA Details
| Gene Symbol: TTC38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017931.2 | NP_060401.2 | 7, 8 | 811 | ||
| GenBank mRNA | AK000706.1 | - | 7, 8 | 767 | ||
| AK300606.1 | - | 6, 7 | 617 | |||
| BC018918.2 | - | 7, 8 | 752 | 4130906 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TTC38| Gene Symbol: TTC38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017931.2 | NP_060401.2 | 7, 8 | 811 | ||
| GenBank mRNA | AK000706.1 | - | 7, 8 | 767 | ||
| AK300606.1 | - | 6, 7 | 617 | |||
| BC018918.2 | - | 7, 8 | 752 | 4130906 | ||