siRNA Details
| Gene Symbol: NRDE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017970.3 | NP_060440.2 | 6, 7 | 1638 | ||
| XM_017021395.1 | XP_016876884.1 | 7, 8 | 1587 | |||
| GenBank mRNA | AK000870.1 | - | 6, 7 | 1436 | ||
| AK024113.1 | - | 1, 2 | 179 | |||
| AK294958.1 | - | 6, 7 | 1442 | |||
| BC098568.1 | - | 6, 7 | 1416 | 30706812 | ||
| BC118673.1 | - | 3, 4 | 971 | 40028659 | ||

NRDE2
Genomic Map