siRNA Details
| Gene Symbol: ZCWPW1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001258008.1 | NP_001244937.1 | 12 | 1502 | ||
| NM_017984.4 | NP_060454.3 | 12 | 1499 | |||
| XM_005250479.2 | XP_005250536.1 | 12 | 1936 | |||
| XM_005250480.2 | XP_005250537.1 | 12 | 1936 | |||
| XM_006716035.3 | XP_006716098.1 | 12 | 1936 | |||
| XM_006716036.2 | XP_006716099.1 | 12 | 1440 | |||
| XM_006716037.3 | XP_006716100.1 | 12 | 1933 | |||
| XM_006716038.3 | XP_006716101.1 | 12 | 1933 | |||
| XM_006716040.3 | XP_006716103.1 | 12 | 1936 | |||
| XM_017012379.1 | XP_016867868.1 | 12 | 1933 | |||
| XM_017012380.1 | XP_016867869.1 | 12 | 1437 | |||
| GenBank mRNA | AK000919.1 | - | 7 | 886 | ||
| AK093038.1 | - | 12 | 1440 | |||
| AK300745.1 | - | 12 | 1358 | |||
| AK302108.1 | - | 11 | 1236 | |||
| AL136735.1 | - | 12 | 1370 | |||
| BC002725.1 | - | 9 | 1200 | 3631266 | ||
| KU178677.1 | - | 7 | 762 | |||
| KU178678.1 | - | 7 | 765 | |||
| KU178679.1 | - | 7 | 762 | |||
| KU178680.1 | - | 7 | 765 | |||
| KU178681.1 | - | 7 | 762 | |||
| KU178682.1 | - | 7 | 762 | |||

ZCWPW1
Genomic Map