siRNA Details
| Gene Symbol: TMEM248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017994.4 | NP_060464.1 | 5 | 908 | ||
| XM_005250482.3 | XP_005250539.1 | 5 | 760 | |||
| GenBank mRNA | AK000961.1 | - | 5 | 894 | ||
| AK222774.1 | - | 5 | 908 | |||
| BC008675.1 | - | 5 | 888 | 3853834 | ||
| BC012562.1 | - | 5 | 903 | 4332094 | ||
| BC041764.2 | - | 5 | 877 | 4152659 | ||

TMEM248
Genomic Map