siRNA Details
| Gene Symbol: TMEM45A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018004.2 | NP_060474.1 | 2 | 428 | ||
| XM_005247569.2 | XP_005247626.1 | 3 | 389 | |||
| XM_017006715.1 | XP_016862204.1 | 3 | 549 | |||
| GenBank mRNA | AB013911.1 | - | 1 | 115 | ||
| AK000996.1 | - | 2 | 428 | |||
| AK129789.1 | - | 2 | 278 | |||
| AY450392.1 | - | 2 | 428 | |||
| BC040355.1 | - | 3 | 362 | 4748104 | ||

TMEM45A
Genomic Map