siRNA Details
| Gene Symbol: GPATCH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018025.2 | NP_060495.2 | 13, 14 | 2205 | ||
| NR_135270.1 | 13, 14 | 2205 | ||||
| GenBank mRNA | AF434677.1 | - | 13, 14 | 1891 | ||
| AK001068.1 | - | 3, 4 | 592 | |||
| AK096005.1 | - | 13, 14 | 1899 | |||
| AK292503.1 | - | 13, 14 | 1899 | |||
| AK315712.1 | - | 13, 14 | 1917 | |||
| AL834465.1 | - | 13, 14 | 1896 | |||
| BC006108.1 | - | 13, 14 | 2205 | 3029084 | ||

GPATCH1
Genomic Map