siRNA Details
Gene Symbol: GPATCH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018025.2 | NP_060495.2 | 1, 2 | 390 | ||
NR_135270.1 | 1, 2 | 390 | ||||
XM_006723255.3 | XP_006723318.1 | 1, 2 | 105 | |||
GenBank mRNA | AF434677.1 | - | 1, 2 | 76 | ||
AK096005.1 | - | 1, 2 | 84 | |||
AK292503.1 | - | 1, 2 | 84 | |||
AK315712.1 | - | 1, 2 | 102 | |||
AL834465.1 | - | 1, 2 | 81 | |||
BC006108.1 | - | 1, 2 | 390 | 3029084 |
