siRNA Details
| Gene Symbol: SAMD4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303614.1 | NP_001290543.1 | 5 | 1256 | ||
| NM_018028.3 | NP_060498.2 | 8 | 1977 | |||
| XM_011527057.2 | XP_011525359.1 | 6 | 1267 | |||
| XM_011527060.2 | XP_011525362.1 | 6 | 2369 | |||
| XM_011527061.2 | XP_011525363.1 | 5 | 1181 | |||
| XM_011527062.2 | XP_011525364.1 | 5 | 1242 | |||
| XM_011527063.2 | XP_011525365.1 | 6 | 1266 | |||
| XM_011527064.2 | XP_011525366.1 | 6 | 1267 | |||
| XM_017026918.1 | XP_016882407.1 | 5 | 1201 | |||
| XM_017026919.1 | XP_016882408.1 | 7 | 1357 | |||
| XM_017026920.1 | XP_016882409.1 | 7 | 1379 | |||
| XM_017026921.1 | XP_016882410.1 | 5 | 1242 | |||
| XM_017026922.1 | XP_016882411.1 | 7 | 1359 | |||
| XM_017026923.1 | XP_016882412.1 | 7 | 6897 | |||
| GenBank mRNA | AK001073.1 | - | 1528 | |||
| BC054518.1 | - | 8 | 1968 | 6501489 | ||
| BC065211.1 | - | 2 | 264 | |||
| BC080641.1 | - | 5 | 1216 | 6645022 | ||
| EF601121.1 | - | 8 | 1964 | |||

SAMD4B
Genomic Map