siRNA Details
| Gene Symbol: AGGF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018046.4 | NP_060516.2 | 7 | 1676 | ||
| GenBank mRNA | AK001145.1 | - | 2 | 188 | ||
| AK289394.1 | - | 3 | 528 | |||
| AY500994.1 | - | 7 | 1595 | |||
| BC015478.1 | - | 2 | 427 | |||
| BC032844.1 | - | 7 | 1662 | 5266978 | ||
| BX648120.1 | - | 3 | 485 | |||

AGGF1
Genomic Map