siRNA Details
| Gene Symbol: TMEM38B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018112.2 | NP_060582.1 | 4, 5 | 677 | ||
| XM_005252075.2 | XP_005252132.1 | 4, 5 | 1124 | |||
| XM_005252076.2 | XP_005252133.1 | 4, 5 | 658 | |||
| XM_005252077.3 | XP_005252134.1 | 4, 5 | 658 | |||
| XM_011518833.2 | XP_011517135.1 | 5, 6 | 809 | |||
| GenBank mRNA | AK001355.1 | - | 4, 5 | 658 | ||
| BC000049.2 | - | 4, 5 | 633 | 3506969 | ||
| CR457258.1 | - | 4, 5 | 541 | |||

TMEM38B
Genomic Map