siRNA Details
| Gene Symbol: APPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001251904.1 | NP_001238833.1 | 14 | 1426 | ||
| NM_001251905.1 | NP_001238834.1 | 14 | 1451 | |||
| NM_018171.3 | NP_060641.2 | 14 | 1408 | |||
| XM_006719472.1 | XP_006719535.1 | 14 | 1345 | |||
| XM_011538530.2 | XP_011536832.1 | 14 | 2878 | |||
| XM_011538531.2 | XP_011536833.1 | 14 | 2541 | |||
| XM_011538532.2 | XP_011536834.1 | 14 | 3867 | |||
| XM_017019551.1 | XP_016875040.1 | 14 | 1813 | |||
| XM_017019552.1 | XP_016875041.1 | 14 | 2523 | |||
| XM_017019553.1 | XP_016875042.1 | 15 | 4121 | |||
| XM_017019554.1 | XP_016875043.1 | 14 | 1327 | |||
| GenBank mRNA | AK290562.1 | - | 14 | 1261 | ||
| AK293783.1 | - | 14 | 1200 | |||
| AK296610.1 | - | 14 | 1207 | |||
| AK297100.1 | - | 14 | 1279 | |||
| AK310377.1 | - | 14 | 1196 | |||
| AY113704.1 | - | 14 | 1408 | |||
| BC033731.1 | - | 14 | 1312 | 5168590 | ||

APPL2
Genomic Map