siRNA Details
| Gene Symbol: PLEKHG6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK001527.1 | - | 10 | 1184 | ||
| BC104751.1 | - | 10 | 1119 | 40035012 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLEKHG6| Gene Symbol: PLEKHG6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK001527.1 | - | 10 | 1184 | ||
| BC104751.1 | - | 10 | 1119 | 40035012 | ||