siRNA Details
| Gene Symbol: LGI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB067503.1 | - | 8 | 1514 | ||
| AF467955.1 | - | 8 | 1522 | |||
| AJ487516.1 | - | 8 | 1522 | |||
| AJ487958.1 | - | 8 | 1544 | |||
| AL833647.1 | - | 7 | 1312 | |||
| BC101759.1 | - | 8 | 1621 | 8069265 | ||
| BC101761.1 | - | 8 | 1621 | 8069267 | ||

LGI2
Genomic Map