siRNA Details
| Gene Symbol: EXD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193360.1 | NP_001180289.1 | 10 | 2096 | ||
| NM_001193361.1 | NP_001180290.1 | 9 | 2012 | |||
| NM_001193362.1 | NP_001180291.1 | 10 | 2255 | |||
| NM_001193363.1 | NP_001180292.1 | 9 | 2171 | |||
| NM_018199.3 | NP_060669.1 | 8 | 1632 | |||
| NR_034164.1 | 11 | 2186 | ||||
| NR_034165.1 | 11 | 2345 | ||||
| XM_005267817.3 | XP_005267874.1 | 8 | 1562 | |||
| XM_011536908.2 | XP_011535210.1 | 8 | 1650 | |||
| XM_017021421.1 | XP_016876910.1 | 9 | 1695 | |||
| GenBank mRNA | AK001600.1 | - | 8 | 1560 | ||
| AK056025.1 | - | 11 | 2132 | |||
| AK292563.1 | - | 10 | 2255 | |||
| AK295601.1 | - | 9 | 2131 | |||
| AL834434.1 | - | 8 | 1554 | |||
| BC001962.1 | - | 8 | 1692 | 3537432 | ||
| BX647767.1 | - | 9 | 1962 | |||
| CR936728.1 | - | 10 | 1971 | |||
| CR936729.1 | - | 9 | 1979 | |||

EXD2
Genomic Map