siRNA Details
| Gene Symbol: TMEM57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018202.5 | NP_060672.2 | 6 | 1226 | ||
| XM_005245931.2 | XP_005245988.1 | 6 | 1187 | |||
| XM_011541704.2 | XP_011540006.1 | 6 | 1260 | |||
| GenBank mRNA | AK001609.1 | - | 6 | 1137 | ||
| AK311472.1 | - | 6 | 1267 | |||
| AY845015.1 | - | 6 | 1156 | |||
| BC032427.1 | - | 6 | 1094 | 5246967 | ||

TMEM57
Genomic Map