siRNA Details
| Gene Symbol: VRTN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018228.2 | NP_060698.2 | 2 | 1221 | ||
| XM_011536911.1 | XP_011535213.1 | 3 | 1268 | |||
| GenBank mRNA | AK001673.1 | - | 2 | 1175 | ||
| AK023213.1 | - | 2 | 1188 | |||
| BC053325.1 | - | 2 | 1204 | 6577658 | ||

VRTN
Genomic Map