siRNA Details
| Gene Symbol: SLC38A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308384.1 | NP_001295313.1 | 3 | 1321 | ||
| NM_018231.2 | NP_060701.1 | 4 | 853 | |||
| XM_017023393.1 | XP_016878882.1 | 3 | 1334 | |||
| XM_017023394.1 | XP_016878883.1 | 4 | 829 | |||
| XM_017023395.1 | XP_016878884.1 | 4 | 829 | |||
| XM_017023396.1 | XP_016878885.1 | 4 | 829 | |||
| XM_017023397.1 | XP_016878886.1 | 4 | 829 | |||
| XM_017023398.1 | XP_016878887.1 | 4 | 829 | |||
| XM_017023399.1 | XP_016878888.1 | 4 | 829 | |||
| GenBank mRNA | AK001677.1 | - | 4 | 846 | ||
| AK022786.1 | - | 4 | 819 | |||
| AK222932.1 | - | 4 | 815 | |||
| AK299424.1 | - | 4 | 478 | |||
| BC001961.1 | - | 3 | 802 | 3510557 | ||
| BC063399.1 | - | 3 | 1321 | |||
| CR457236.1 | - | 2 | 453 | |||

SLC38A7
Genomic Map