siRNA Details
| Gene Symbol: KIRREL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286349.1 | NP_001273278.1 | 6, 7 | 1147 | ||
| NM_018240.6 | NP_060710.3 | 8, 9 | 1447 | |||
| GenBank mRNA | AK001707.1 | - | 1, 2 | 139 | ||
| AK297788.1 | - | 6, 7 | 775 | |||
| AY017369.1 | - | 8, 9 | 1043 | |||
| AY302131.1 | - | 6, 7 | 1142 | |||
| BC051356.1 | - | 1, 2 | 110 | |||
| BC067097.1 | - | 1, 2 | 110 | |||
| BC082969.1 | - | 1, 2 | 110 | |||
| BC109192.1 | - | 8, 9 | 1110 | 40001501 | ||
| BC109193.1 | - | 8, 9 | 1110 | 40001502 | ||

KIRREL
Genomic Map