siRNA Details
| Gene Symbol: SLC47A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018242.2 | NP_060712.2 | 13 | 1218 | ||
| GenBank mRNA | AK001709.1 | - | 13 | 1218 | ||
| AK222625.1 | - | 13 | 1222 | |||
| AK293196.1 | - | 6 | 511 | |||
| AK298254.1 | - | 4 | 460 | |||
| AK301854.1 | - | 12 | 1175 | |||
| AK304647.1 | - | 5 | 397 | |||
| AK310270.1 | - | 11 | 897 | |||
| BC010661.2 | - | 13 | 1207 | 3874616 | ||
| BC050592.1 | - | 13 | 1192 | |||
| BC058882.1 | - | 10 | 1094 | 5188651 | ||

SLC47A1
Genomic Map