siRNA Details
| Gene Symbol: SLC47A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018242.2 | NP_060712.2 | 5, 6 | 583 | ||
| GenBank mRNA | AK001709.1 | - | 5, 6 | 583 | ||
| AK222625.1 | - | 5, 6 | 587 | |||
| AK301854.1 | - | 4, 5 | 540 | |||
| AK302620.1 | - | 5, 6 | 583 | |||
| AK310270.1 | - | 5, 6 | 402 | |||
| BC010661.2 | - | 5, 6 | 572 | 3874616 | ||
| BC050592.1 | - | 5, 6 | 557 | |||
| BC058882.1 | - | 5, 6 | 559 | 5188651 | ||

SLC47A1
Genomic Map