siRNA Details
| Gene Symbol: WDR41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018268.3 | NP_060738.2 | 2 | 417 | ||
| XM_005248551.4 | XP_005248608.1 | 2 | 384 | |||
| XM_005248552.3 | XP_005248609.1 | 2 | 384 | |||
| XM_011543505.2 | XP_011541807.1 | 2 | 179 | |||
| XM_011543507.2 | XP_011541809.1 | 2 | 384 | |||
| XM_017009606.1 | XP_016865095.1 | 2 | 827 | |||
| GenBank mRNA | AF115511.1 | - | 2 | 166 | ||
| AK001766.1 | - | 2 | 363 | |||
| AK021855.1 | - | 2 | 203 | |||
| AK023977.1 | - | 2 | 119 | |||
| AK293316.1 | - | 2 | 76 | |||
| AK295249.1 | - | 2 | 97 | |||
| AK296423.1 | - | 2 | 193 | |||
| AK304329.1 | - | 2 | 99 | |||
| BC040241.1 | - | 2 | 169 | 3447269 | ||
| BM818066.1 | - | 2 | 384 | |||
| DC403958.1 | - | 2 | 418 | |||
| KU178690.1 | - | 2 | 97 | |||
| KU178691.1 | - | 2 | 97 | |||

WDR41
Genomic Map