siRNA Details
| Gene Symbol: MRGBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018270.5 | NP_060740.1 | 2 | 365 | ||
| NR_136405.1 | 2 | 365 | ||||
| GenBank mRNA | AB085682.1 | - | 2 | 265 | ||
| AK001776.1 | - | 2 | 317 | |||
| BC009889.2 | - | 2 | 284 | 4026378 | ||
| BC014235.2 | - | 2 | 289 | 4555060 | ||
| BC018841.2 | - | 2 | 284 | 3050866 | ||
| BM920599.1 | - | 271 | ||||

MRGBP
Genomic Map