siRNA Details
| Gene Symbol: TMEM143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303538.1 | NP_001290467.1 | 4, 5 | 917 | ||
| NM_001303539.1 | NP_001290468.1 | 4, 5 | 827 | |||
| NM_001303540.1 | NP_001290469.1 | 3, 4 | 722 | |||
| NM_018273.3 | NP_060743.2 | 5, 6 | 1022 | |||
| NR_130317.1 | 3, 4 | 676 | ||||
| GenBank mRNA | AK129801.1 | - | 5, 6 | 725 | ||
| AK291521.1 | - | 3, 4 | 423 | |||
| AK297591.1 | - | 3, 4 | 377 | |||
| AK297610.1 | - | 4, 5 | 618 | |||
| AK298317.1 | - | 4, 5 | 528 | |||
| AK303635.1 | - | 4 | 1041 | |||
| AY358161.1 | - | 3, 4 | 414 | |||
| BC016919.1 | - | 5, 6 | 714 | 4400601 | ||
| BC040021.1 | - | 5, 6 | 720 | |||

TMEM143
Genomic Map