siRNA Details
| Gene Symbol: C7orf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303470.1 | NP_001290399.1 | 7 | 740 | ||
| NM_018275.4 | NP_060745.3 | 7 | 1240 | |||
| GenBank mRNA | AK001787.1 | - | 3 | 220 | ||
| AK054872.1 | - | 7 | 736 | |||
| AK302342.1 | - | 5 | 588 | |||
| BC002749.2 | - | 7 | 640 | |||
| BC015722.2 | - | 7 | 1198 | 3920412 | ||

C7orf43
Genomic Map