siRNA Details
| Gene Symbol: C7orf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018275.4 | NP_060745.3 | 1 | 240 | ||
| GenBank mRNA | AK310170.1 | - | 1 | 221 | ||
| BC015722.2 | - | 1 | 198 | 3920412 | ||
| HY066862.1 | - | 1 | 240 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C7orf43| Gene Symbol: C7orf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018275.4 | NP_060745.3 | 1 | 240 | ||
| GenBank mRNA | AK310170.1 | - | 1 | 221 | ||
| BC015722.2 | - | 1 | 198 | 3920412 | ||
| HY066862.1 | - | 1 | 240 | |||