siRNA Details
| Gene Symbol: TMEM19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018279.3 | NP_060749.2 | 1 | 659 | ||
| GenBank mRNA | AK001798.1 | - | 1 | 261 | ||
| AK223148.1 | - | 1 | 257 | |||
| AK315587.1 | - | 1 | 218 | |||
| BC008596.1 | - | 1 | 659 | 4154303 | ||

TMEM19
Genomic Map