siRNA Details
| Gene Symbol: TMEM19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018279.3 | NP_060749.2 | 3, 4 | 971 | ||
| GenBank mRNA | AK001798.1 | - | 3, 4 | 573 | ||
| AK223148.1 | - | 3, 4 | 569 | |||
| AK315587.1 | - | 3, 4 | 530 | |||
| BC008596.1 | - | 3, 4 | 971 | 4154303 | ||

TMEM19
Genomic Map