siRNA Details
| Gene Symbol: RBM41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171080.1 | NP_001164551.1 | 5 | 679 | ||
| NM_001324242.1 | NP_001311171.1 | 6 | 751 | |||
| NM_001324243.1 | NP_001311172.1 | 6 | 844 | |||
| NM_001324244.1 | NP_001311173.1 | 6 | 751 | |||
| NM_018301.4 | NP_060771.3 | 5 | 679 | |||
| XM_011530980.2 | XP_011529282.1 | 6 | 843 | |||
| XM_011530981.2 | XP_011529283.1 | 5 | 771 | |||
| GenBank mRNA | AK001878.1 | - | 5 | 652 | ||
| BC006986.1 | - | 5 | 656 | 3995903 | ||

RBM41
Genomic Map